missing translation for 'onlineSavingsMsg'
Learn More

Skatole, MedChemExpress

Product Code. 16429326
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
missing translation for 'unitSize'
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16429326 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 16429326 missing translation for 'mfr' Medchem Express missing translation for 'supplierNo' HYW007355

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Skatole is produced by intestinal bacteria, regulates intestinal epithelial cellular functions through activating aryl hydrocarbon receptors and p38.

Skatole is produced by intestinal bacteria, regulates intestinal epithelial cellular functions through activating aryl hydrocarbon receptors and p38.

Specifications

Chemical Name or Material Skatole
CAS 83-34-1
Color White
Purity Grade Notes Research
Molecular Formula C9H9N
Quantity 100 mg
Synonym 3-Methylindole 3-Methyl-1H-indole
Solubility Information DMSO : 100 mg/mL (762.37 mM; Need ultrasonic)
SMILES CC1=CNC2=CC=CC=C12
Molecular Weight (g/mol) 131.17
Formula Weight 131.17
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.