Learn More
Manutan Skate wheel Expandaveyor, Expandaveyor wheels
Product Code. 12810279
Encompass_Preferred
Product Code. 12810279
Brand: Manutan A341372
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A highly versatile conveyor system for use alone or to supplement existing installations in stores, warehouses, assembly lines and loading/unloading vBright zinc plated steel support frames and lattice straps with plastic roller bearings or skate wheels. Adjustable telescopic legs fitted at every 1. 5m of expanded length. O/A width: track width plus 62mm. Pitch: 125mm when expanded. Plastic roller bearings - 50mm diameter shock proof, rigid PVC with double ball race and ball bearings. Plastic skate wheels - 48mm diameter shock proof PVC with double ball race, and steel axles with PVC covers. Adjustable support legs - All fitted with 100mm diameter braked castors. 125kg capacity Expandaveyor support legs have additional lattice knee bracing. Height 650-1000mm.
  • Track width: 500mm
  • Expanded length: 3.5m
  • Closed length: 1.00m