missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SKAR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17466-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein that interacts with the DNA polymerase delta p50 subunit. This protein is a specific target of S6 kinase 1 and regulates cell growth. Two transcript variants that encode different protein isoforms have been identified.Specifications
| SKAR | |
| Polyclonal | |
| Unconjugated | |
| POLDIP3 | |
| KIAA1649, p46, PDIP46, POLDIP3, S6K1 Aly/REF like target, SKAR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 315170, 73826, 84271 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BG81, Q9BY77 | |
| Poldip3 | |
| SKAR Fusion Protein Ag11338 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction