missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SKAP55 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14724-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the vaccinia-related kinase (VRK) family of serine/threonine protein kinases. This gene is widely expressed in human tissues and has increased expression in actively dividing cells, such as those in testis, thymus, fetal liver, and carcinomas. Its protein localizes to the nucleus and has been shown to promote the stability and nuclear accumulation of a transcriptionally active p53 molecule and, in vitro, to phosphorylate Thr18 of p53 and reduce p53 ubiquitination. This gene, therefore, may regulate cell proliferation. This protein also phosphorylates histone, casein, and the transcription factors ATF2 (activating transcription factor 2) and c-JUN.Specifications
| SKAP55 | |
| Polyclonal | |
| Unconjugated | |
| SKAP1 | |
| pp55, SCAP1, SKAP 55, SKAP1, SKAP55 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 286975, 78473, 8631 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q3UUV5, Q4V7G1, Q86WV1 | |
| Skap1 | |
| SKAP55 Fusion Protein Ag6467 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction