missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SKAP55 Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA5148824

Product Code. 30533894

  • £415.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

This gene encodes a T cell adaptor protein, a class of intracellular molecules with modular domains capable of recruiting additional proteins but that exhibit no intrinsic enzymatic activity. The encoded protein contains a unique N-terminal region followed by a PH domain and C-terminal SH3 domain. Along with the adhesion and degranulation-promoting adaptor protein, the encoded protein plays a critical role in inside-out signaling by coupling T-cell antigen receptor stimulation to the activation of integrins.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SKAP55
Polyclonal
Unconjugated
SKAP1
1700091G21Rik; epididymis secretory sperm binding protein Li 81p; HEL-S-81p; ortholog of human src family associated phosphoprotein 1 SCAP1; pp55; SCAP1; Skap1; Skap55; Skap-55; SKAP55 adapter protein; SKAP55 adaptor protein; src family associated phosphoprotein 1; src family-associated phosphoprotein 1; src kinase associated phosphoprotein 1; src kinase-associated phosphoprotein 1; Src kinase-associated phosphoprotein of 55 kDa
Goat
Antigen affinity chromatography
RUO
8631
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Western Blot
0.2 mg/mL
PBS with trehalose and no preservative
Q86WV1
SKAP1
E. coli-derived recombinant human SKAP55, Met1-Arg358.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.