missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SKAP2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518014
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Fyb (Fyn binding protein) and the anchoring proteins SKAP55 and SKAP55-R (SKAP55-related protein) associate with the tyrosine kinase p59fyn. SKAP55 and SKAP55-R bind to Fyb through their SH3 domains and function as substrates for p59Fyn in resting T cells. SKAP55 contains an N-terminal pleckstrin homology domain and a C-terminal SH3 domain binding motif of adjacent arginine and lysine residues followed by tandem tyrosines (i.e. RKxxYxxY). SKAP55-R, similar in overall structure to SKAP55, contains a coiled-coil N-terminal domain. SKAP55 associates with SLAP-130, another component of the Fyn complex, which plays a role in the regulation of signaling events initiated by lymphocyte antigen receptors leading up to T cell activation. The human SKAP55 gene maps to chromosome 17q21.32 and encodes a 359 amino acid protein.
Specifications
| SKAP2 | |
| Polyclonal | |
| Unconjugated | |
| SKAP2 | |
| 2610021A10Rik; AA960083; BB137539; Fyn-associated phosphoprotein SKAP55 homologue; MGC10411; MGC33304; mSKAP55R; PRAP; pyk2/RAFTK-associated protein; RA70; Retinoic acid-induced protein 70; Saps; SCAP2; Scap55r; Skap2; SKAP55 homolog; SKAP55 homologue; SKAP-55HOM; SKAP55-HOM; SKAP55R; SKAP-HOM; src family associated phosphoprotein 2; src family-associated phosphoprotein 2; src kinase associated phosphoprotein 2; src kinase-associated phosphoprotein 2; Src kinase-associated phosphoprotein 55-related protein; src kinase-associated phosphoprotein of 55-related protein; src-associated adapter protein with PH and SH3 domains; Src-associated adaptor protein | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 8935 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O75563 | |
| SKAP2 | |
| Synthetic peptide sequence (GLVPKAYIMEMYDI) corresponding to the C-terminus amino acids of SKAP2 (aa 346-359). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction