missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SKAP2 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2G3G9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66367-1-IG-20UL
This item is not returnable.
View return policy
Description
Fyb (Fyn binding protein) and the anchoring proteins SKAP55 and SKAP55-R (SKAP55-related protein) associate with the tyrosine kinase p59fyn. SKAP55 and SKAP55-R bind to Fyb through their SH3 domains and function as substrates for p59Fyn in resting T cells. SKAP55 contains an N-terminal pleckstrin homology domain and a C-terminal SH3 domain binding motif of adjacent arginine and lysine residues followed by tandem tyrosines (i.e. RKxxYxxY). SKAP55-R, similar in overall structure to SKAP55, contains a coiled-coil N-terminal domain. SKAP55 associates with SLAP-130, another component of the Fyn complex, which plays a role in the regulation of signaling events initiated by lymphocyte antigen receptors leading up to T cell activation. The human SKAP55 gene maps to chromosome 17q21.32 and encodes a 359 amino acid protein.Specifications
| SKAP2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75563, Q3UND0, Q920G0 | |
| Skap2 | |
| SKAP2 Fusion Protein Ag3615 | |
| 20 μL | |
| Primary | |
| Pig, Mouse, Rat, Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
| 2G3G9 | |
| Unconjugated | |
| SKAP2 | |
| PRAP, Pyk2/RAFTK associated protein, RA70, SAPS, SCAP2, SKAP 55HOM, SKAP HOM, SKAP2, SKAP55 homolog, SKAP55R | |
| Mouse | |
| Protein G | |
| RUO | |
| 100625903, 155183, 54353, 8935 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction