missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SKAP Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £364.00
Specifications
| Antigen | SKAP |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
SKAP Polyclonal antibody specifically detects SKAP in Human samples. It is validated for Western BlotSpecifications
| SKAP | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| C15orf23, chromosome 15 open reading frame 23, FLJ14502, HSD11, MGC141728, MGC141729, putative TRAF4-associated factor 1, SKAP, small kinetochore associated protein, TRAF4 associated factor 1, TRAF4AF1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human KNSTRN (NP_001136233.1). MAAPEAPPLDRVFRTTWLSTECDSHPLPPSYRKFLFETQAADLAGGTTVAAGNLLNESEKDCGQDRRAPGVQPCRLVTMTSVVKTVYSLQPPSALSGGQPADTQTRATSKSLLPVRSKEVDVSKQLHSGGPENDVTKITKLRRENGQMKATDTATRRNVRKGYKPLSKQKSEEELKDKNQLLEAVNKQLHQKLTETQGELKDLTQKVELLEKFRDNCLAILESKGLDPALGSETLASRQESTTDHMDSMLLLETLQEELKLFNETAKKQM | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 90417 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title