missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SKA3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5139853

Product Code. 17844370

  • £388.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: RTSLVLNSDT CFENLTDPSS PTISSYENLL RTPTPPEVTK IPEDILQLLS KYNSNLATPI AIKAVPPSKR FLKHGQNIRD VSNKEN Highest antigen sequence indentity to the following orthologs: Rat - 58%, Mouse - 45%.

Upon entry into mitosis, the cell's microtubule (MT) network forms the mitotic spindle, allowing the segregation of paired chromosomes. Proteinaceous structures on centromeric chromatin termed kinetochores (KT) are essential for the proper attachment of the chromosomes to the spindle MTs. A recently discovered spindle and kinetochore complex, comprised of proteins SKA1, SKA2, and SKA3, has been found to be required for stable KT-MT interactions and timely anaphase onset. Like with SKA1 or SKA2, depletion of SKA3 by siRNA delays anaphase transition, resulting in a prolonged a metaphase-like state. These SKA3-depleted cells accumulate high levels of the checkpoint protein Bub1 at kinetochores, suggesting the SKA complex plays a key role in spindle checkpoint silencing and the maintenance of chromosome cohesion in mitosis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SKA3
Polyclonal
Unconjugated
SKA3
C13orf3; F630043A04Rik; RAMA1; RGD1307201; RP11-101P17.12-002; Ska3; spindle and kinetochore associated complex subunit 3; spindle and kinetochore-associated protein 3
Rabbit
Antigen affinity chromatography
RUO
221150
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8IX90
SKA3
Recombinant protein corresponding to Human SKA3. Recombinant protein control fragment (Product #RP-109000).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.