missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SKA3 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76311PCP
This item is not returnable.
View return policy
Description
SKA3 Polyclonal antibody specifically detects SKA3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| SKA3 | |
| Polyclonal | |
| Western Blot, ELISA | |
| spindle and kinetochore associated complex subunit 3, spindle and kinetochore-associated protein 3 | |
| Antibody was raised against a 13 amino acid synthetic peptide from near the carboxy terminus of human SKA3. The immunogen is located within amino acids 290 - 340 of SKA3. Amino Acid Squence: STNYPLSKTNSSS | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| PerCP | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 221150 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction