missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SKA3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76311
This item is not returnable.
View return policy
Description
SKA3 Polyclonal specifically detects SKA3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| SKA3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| spindle and kinetochore associated complex subunit 3, spindle and kinetochore-associated protein 3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 221150 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000 | |
| NP_659498 | |
| SKA3 | |
| Antibody was raised against a 13 amino acid synthetic peptide from near the carboxy terminus of human SKA3. The immunogen is located within amino acids 290 - 340 of SKA3. Amino Acid Squence: STNYPLSKTNSSS | |
| 0.1 mg | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction