missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SKA 31

Description
SKA 31 is an activator of KCa3.1 and KCa2 channels (EC50 values are 260, 2900, 2900 nM for KCa3.1, KCa2.1 and KCa2.2 respectively). Potentiates acetylcholine-induced EDHF-type dilations of mouse carotid arteries and lowers blood pressure in normotensive and hypertensive mice.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | SKA 31 |
| Purity | >99% |
| Molecular Weight (g/mol) | 200.26 |
| Inhibitors | SKA 31 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?