missing translation for 'onlineSavingsMsg'
Learn More

SKA-31, MedChemExpress

Product Code. 16467227
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467227 5 mg 5mg
16435376 100 mg 100mg
16424518 10 mg 10mg
16410827 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16467227 Supplier Medchem Express Supplier No. HY111655

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SKA-31 is a potent potassium channel activator with EC50s of 260 nM, 1.9 μM, 2.9 μM, and 2.9 μM for KCa3.1, KCa2.2, KCa2.1 and KCa2.3, respectively. SKA-31 potentiates endothelium-derived hyperpolarizing factor response and lowers blood pressure.

SKA-31 is a potent potassium channel activator with EC50s of 260 nM, 1.9 μM, 2.9 μM, and 2.9 μM for KCa3.1, KCa2.2, KCa2.1 and KCa2.3, respectively. SKA-31 potentiates endothelium-derived hyperpolarizing factor response and lowers blood pressure.

Specifications

Chemical Name or Material SKA-31
Physical Form Solid
CAS 40172-65-4
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C11H8N2S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 150 mg/mL (749.03 mM; Need ultrasonic)
SMILES NC1=NC2=C3C=CC=CC3=CC=C2S1
Molecular Weight (g/mol) 200.26
Formula Weight 200.26
Percent Purity 99.45%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.