missing translation for 'onlineSavingsMsg'
Learn More

Tocris™ SK 609

Product Code. p-7199095
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18749446

Brand: Tocris™ 7039/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dopamine D3 receptor biased agonist

SK 609 is a dopamine D3 receptor biased agonist. Preferentially signals via the G protein-dependent pathway (EC50 values for G protein and β-arrestin-dependent signaling are 1.1 and 50.2 μM, respectively). Exhibits selectivity for D3 over D2 receptors. Improves motor deficits in rodent Parkinson's disease models and ameliorates L-DOPA (Cat. No. 3788) induced abnormal involuntary movements.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-chloro-α-methyl-benzenepropanamine hydrochloride
CAS 1092797-77-7
Quantity 25 mg
Target Dopamine D3 Receptor Agonists
Molecular Formula C10H14ClN.HCl
Formula Weight 3333
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.