missing translation for 'onlineSavingsMsg'
Learn More

Tocris™ SK 609

Product Code. 18749446
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18749446 25 mg 25mg
18729636 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18749446 Supplier Tocris™ Supplier No. 7039/25
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Dopamine D3 receptor biased agonist

SK 609 is a dopamine D3 receptor biased agonist. Preferentially signals via the G protein-dependent pathway (EC50 values for G protein and β-arrestin-dependent signaling are 1.1 and 50.2 μM, respectively). Exhibits selectivity for D3 over D2 receptors. Improves motor deficits in rodent Parkinson's disease models and ameliorates L-DOPA (Cat. No. 3788) induced abnormal involuntary movements.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-chloro-α-methyl-benzenepropanamine hydrochloride
CAS 1092797-77-7
Quantity 25 mg
Target Dopamine D3 Receptor Agonists
Molecular Formula C10H14ClN.HCl
Formula Weight 3333
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.