missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SJFa SJFalpha SJFalpha
GREENER_CHOICE

Product Code. 18758416
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18758416 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18758416 Supplier Tocris Bioscience™ Supplier No. 7268/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective p38α PROTAC™

SJFα is a potent and selective p38α PROTAC™ Degrader (DC50 = 7.16 nM and Dmax = 97.4%). SJFα comprises the multikinase inhibitor foretinib joined by a linker to a VHL ligand. Displays significantly lower potency degradation at p38δ (DC50 = 299 nM). Exhibits no significant degradation of p38β or γ. or related MAPKs, ERK1/2, or JNK1/2. PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(3-Fluoro-4-((7-(4-(4-(2-((-1-((2 S,4 R)-4-hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-2-oxoethoxy)butoxy)butoxy)-6-methoxyquinolin-4-yl)oxy)phenyl)-N-(4-fluorophenyl)cyclopropane-1,1-dicarb
CAS 2254609-27-1
Quantity 5 mg
Target Active Degraders
Molecular Formula C59H67F2N7O11S
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.