missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SJF 1521

Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
SJF 1521 is a selective EGFR PROTAC™ Degrader. SJF 1521 comprises the EGFR inhibitor lapatinib (Cat. No. 6811) joined by a linker to a von Hippel-Lindau (VHL) recruiting ligand. Exhibits selectivity for EGFR, including mutant forms, over HER2. Induces degradation of EGFR in OVCAR8 cells. PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | (2 S,4 R)-1-(-2-(tert-Butyl)-14-(4-(4-((3-chloro-4-((3-fluorobenzyl)oxy)phenyl)amino)quinazolin-6-yl)phenoxy)-4-oxo-6,9,12-trioxa-3-azatetradecanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide |
| CAS | 2230821-40-4 |
| Quantity | 5 mg |
| Target | Active Degraders |
| Molecular Formula | C57H61ClFN7O9S |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction