missing translation for 'onlineSavingsMsg'
Learn More

SJ000291942, MedChemExpress

Product Code. 16475466
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16475466

Brand: Medchem Express HY112331

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SJ000291942 is an activator of the canonical bone morphogenetic proteins (BMP) signaling pathway. BMPs are members of the transforming growth factor beta (TGFβ) family of secreted signaling molecules.

SJ000291942 is an activator of the canonical bone morphogenetic proteins (BMP) signaling pathway. BMPs are members of the transforming growth factor beta (TGFβ) family of secreted signaling molecules.

Specifications

Chemical Name or Material SJ000291942
CAS 425613-09-8
Color White
Purity Grade Notes Research
Molecular Formula C16H15FN2O4
Quantity 100 mg
Solubility Information DMSO : ≥ 150 mg/mL (471.25 mM)
SMILES O=C(NC1=CC=C(F)C([N+]([O-])=O)=C1)COC2=CC=C(CC)C=C2
Molecular Weight (g/mol) 318.3
Formula Weight 318.3
Percent Purity 98.41%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.