missing translation for 'onlineSavingsMsg'
Learn More

SJ-172550, MedChemExpress

Product Code. 16477108
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477108 10 mg 10mg
16463147 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16477108 Supplier Medchem Express Supplier No. HY16664
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress SJ-172550 is a small molecule inhibitor of MDMX; competes for the wild type p53 peptide binding to MDMX with an EC50 of 5 μM.

SJ-172550 is a small molecule inhibitor of MDMX

Specifications

Chemical Name or Material SJ-172550
CAS 431979-47-4
Color Red
Purity Grade Notes Research
Molecular Formula C22H21ClN2O5
Quantity 10 mg
Solubility Information DMSO : 33.33 mg/mL (77.72 mM; Need ultrasonic)
SMILES O=C(/C1=C/C2=CC(Cl)=C(OCC(OC)=O)C(OCC)=C2)N(N=C1C)C3=CC=CC=C3
Molecular Weight (g/mol) 428.87
Formula Weight 428.87
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.