missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIX6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599307
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SIX6.
The protein encoded by this gene is a homeobox protein that is similar to the Drosophila 'sine oculis' gene product. This gene is found in a cluster of related genes on chromosome 14 and is thought to be involved in eye development. Defects in this gene are a cause of isolated microphthalmia with cataract type 2 (MCOPCT2).
Specifications
| SIX6 | |
| Polyclonal | |
| Unconjugated | |
| SIX6 | |
| Homeobox protein SIX6; homeodomain protein OPTX2; MCOPCT2; ODRMD; optic homeobox 2; Optx2; sine oculis homeobox 6; sine oculis homeobox homolog 6; sine oculis homeobox protein 6; sine oculis homeobox-like protein 6; sine oculis-related homeobox 6; sine oculis-related homeobox 6 homolog; SIX homeobox 6; Six6; SIX9; Six9 protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20476, 314221, 4990 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95475, Q9QZ28 | |
| SIX6 | |
| A synthesized peptide derived from human SIX6(Accession O95475), corresponding to amino acid residues Q130-R180. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction