missing translation for 'onlineSavingsMsg'
Learn More

SIX3 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Product Code. 18300305 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18300305 25 μg 25µL
18331604 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18300305 Supplier Novus Biologicals Supplier No. NBP31042125UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SIX3 Polyclonal specifically detects SIX3 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIX3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS buffer, 2% sucrose
Gene Alias holoprosencephaly 2, alobar or semilobar, homeobox protein SIX3, HPE2, Sine oculis homeobox homolog 3, sine oculis homeobox homolog 3 (Drosophila), SIX homeobox 3
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C terminal region of mouse SIX3 (NP_035511). Peptide sequence MRSLAEPGCPTHGSAESPSTAASPTTSVSSLTERADTGTSILSVTSSDSE
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6496
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.