missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIX3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103901
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SIX3.
This gene encodes a member of the sine oculis homeobox transcription factor family. The encoded protein plays a role in eye development. Mutations in this gene have been associated with holoprosencephaly type 2.
Specifications
| SIX3 | |
| Polyclonal | |
| Unconjugated | |
| Six3 | |
| E130112M24Rik; Homeobox protein SIX3; HPE2; sine oculis homeobox 3-like protein; sine oculis homeobox homolog 3; sine oculis homeobox-like protein 3; sine oculis-related homeobox 3; sine oculis-related homeobox 3 homolog; SIX homeobox 3; Six3; Six3a; Six3alpha; Six3b; Six3beta | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 20473, 6496, 78974 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95343, Q62233 | |
| Six3 | |
| A synthesized peptide derived from human SIX3(Accession O95343), corresponding to amino acid residues R269-S319. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction