missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584322
This item is not returnable.
View return policy
Description
Immunogen sequence: ADPLQHHHGL QDSILNPMSA NLVDLGS Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 96%.
This gene is a member of the vertebrate gene family which encode proteins homologous to the Drosophila 'sine oculis' homeobox protein. SIX2 is a transcription factor which, like other members of this gene family, may be involved in limb or eye development.
Specifications
| SIX2 | |
| Polyclonal | |
| Unconjugated | |
| SIX2 | |
| homeobox protein SIX2; Sine oculis homeobox homolog 2; sine oculis-related homeobox 2; sine oculis-related homeobox 2 homolog; SIX homeobox 2; SIX2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10736 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NPC8 | |
| SIX2 | |
| Recombinant protein corresponding to Human SIX2. Recombinant protein control fragment (Product #RP-106741). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction