missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIX1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101643
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SIX1.
The protein encoded by this gene is a homeobox protein that is similar to the Drosophila 'sine oculis' gene product. This gene is found in a cluster of related genes on chromosome 14 and is thought to be involved in limb development. Defects in this gene are a cause of autosomal dominant deafness type 23 (DFNA23) and branchiootic syndrome type 3 (BOS3).
Specifications
| SIX1 | |
| Polyclonal | |
| Unconjugated | |
| SIX1 | |
| BB138287; BOS3; DFNA23; Homeobox protein SIX1; sine oculis homeobox homolog 1; sine oculis-related homeobox 1; sine oculis-related homeobox 1 homolog; SIX homeobox 1; SIX1; TIP39 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 20471, 6495 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q15475, Q62231 | |
| SIX1 | |
| A synthesized peptide derived from human SIX1(Accession Q15475), corresponding to amino acid residues V111-T161. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction