missing translation for 'onlineSavingsMsg'
Learn More

Sivelestat sodium tetrahydrate, MedChemExpress

Product Code. 16437316
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16437316 100 mg 100mg
16463367 50 mg 50mg
16447348 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16437316 Supplier Medchem Express Supplier No. HY17443B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sivelestat (EI546) sodium tetrahydrate is a competitive inhibitor of human neutrophil elastase, with an IC50 of 44 nM and a Ki of 200 nM. Sivelestat (EI546) sodium tetrahydrate has the potential for the study of acute lung injury/acute respiratory distress syndrome or disseminated intravascular coagulation in COVID-19.

Sivelestat (EI546) sodium tetrahydrate is a competitive inhibitor of human neutrophil elastase, with an IC50 of 44 nM and a Ki of 200 nM. Sivelestat (EI546) sodium tetrahydrate has the potential for the study of acute lung injury/acute respiratory distress syndrome or disseminated intravascular coagulation in COVID-19.

Specifications

Chemical Name or Material Sivelestat sodium tetrahydrate
CAS 201677-61-4
Color White
Purity Grade Notes Research
Molecular Formula C20H29N2NaO11S
Quantity 100 mg
Synonym EI546 sodium tetrahydrateLY544349 sodium tetrahydrateONO5046 sodium tetrahydrate
Solubility Information DMSO : 250 mg/mL (473.03 mM; Need ultrasonic)
SMILES CC(C)(C)C(OC1=CC=C(S(=O)(NC2=CC=CC=C2C(NCC(O[Na])=O)=O)=O)C=C1)=O.O.O.O.O
Molecular Weight (g/mol) 528.51
Formula Weight 528.51
Percent Purity 99.06%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.