missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sivelestat sodium, MedChemExpress
Description
Sivelestat (EI546) sodium is a competitive inhibitor of human neutrophil elastase, with an IC50 of 44 nM and a Ki of 200 nM. Sivelestat (EI546) sodium has the potential for the study of acute lung injury/acute respiratory distress syndrome or disseminated intravascular coagulation in COVID-19.
Specifications
Specifications
| Chemical Name or Material | Sivelestat sodium |
| CAS | 150374-95-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C20H21N2NaO7S |
| Quantity | 50 mg |
| Synonym | ONO5046-Na Sodium sivelestat EI546 sodium LY544349 sodium |
| Solubility Information | DMSO : 100 mg/mL (219.09 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.19 mM; ultrasonic and warming and heat to 80°C) |
| SMILES | CC(C)(C)C(OC1=CC=C(S(=O)(NC2=CC=CC=C2C(NCC(O[Na])=O)=O)=O)C=C1)=O |
| Molecular Weight (g/mol) | 456.44 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?