missing translation for 'onlineSavingsMsg'
Learn More

Sivelestat sodium, MedChemExpress

Product Code. 16437348
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16437348 10 mg 10mg
16453367 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16437348 Supplier Medchem Express Supplier No. HY17443A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sivelestat (EI546) sodium is a competitive inhibitor of human neutrophil elastase, with an IC50 of 44 nM and a Ki of 200 nM. Sivelestat (EI546) sodium has the potential for the study of acute lung injury/acute respiratory distress syndrome or disseminated intravascular coagulation in COVID-19.

Sivelestat (EI546) sodium is a competitive inhibitor of human neutrophil elastase, with an IC50 of 44 nM and a Ki of 200 nM. Sivelestat (EI546) sodium has the potential for the study of acute lung injury/acute respiratory distress syndrome or disseminated intravascular coagulation in COVID-19.

Specifications

Chemical Name or Material Sivelestat sodium
CAS 150374-95-1
Color White
Purity Grade Notes Research
Molecular Formula C20H21N2NaO7S
Quantity 10 mg
Synonym ONO5046-Na Sodium sivelestat EI546 sodium LY544349 sodium
Solubility Information DMSO : 100 mg/mL (219.09 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.19 mM; ultrasonic and warming and heat to 80°C)
SMILES CC(C)(C)C(OC1=CC=C(S(=O)(NC2=CC=CC=C2C(NCC(O[Na])=O)=O)=O)C=C1)=O
Molecular Weight (g/mol) 456.44
Formula Weight 456.44
Percent Purity 99.13%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.