missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIVA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566771
This item is not returnable.
View return policy
Description
Immunogen sequence: PLQLKVRVSQR ELSRGVCAER YSQEVFEKTK RLLFLGAQAY LDHVWDEGCA VVHLPESPKP GPTGAPRAAR GQMLIGPDGR LI Highest antigen sequence identity to the following orthologs - mouse 69%, rat 76%.
Induces CD27-mediated apoptosis. Inhibits BCL2L1 isoform Bcl-x(L) anti-apoptotic activity. Inhibits activation of NF-kappa-B and promotes T-cell receptor-mediated apoptosis.
Specifications
| SIVA1 | |
| Polyclonal | |
| Unconjugated | |
| SIVA1 | |
| apoptosis regulatory protein Siva; Cd27 binding protein; CD27-binding (Siva) protein; CD27-binding protein; CD27BP; SIVA; SIVA1; Siva-1; SIVA1 apoptosis inducing factor; SIVA1, apoptosis-inducing factor; Siva-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10572 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15304 | |
| SIVA1 | |
| Recombinant Human SIVA1. Recombinant protein control fragment (Product #RP-107428). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction