missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIVA Antibody (OTI2F8) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74208
This item is not returnable.
View return policy
Description
SIVA Monoclonal specifically detects SIVA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| SIVA | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
| CD27-binding (Siva) protein, CD27BPCD27-binding protein, Siva-1, SIVA1, apoptosis-inducing factor, Siva-2, SIVAapoptosis regulatory protein Siva | |
| Mouse | |
| 18.5 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI2F8 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| SIVA1 | |
| Full length human recombinant protein of human SIVA1(NP_006418) produced in HEK293T cell. | |
| Affinity Purified | |
| Apoptosis | |
| 10572 | |
| Human | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction