missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIVA Antibody, Alexa Fluor™ 532, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP2-41150AF532
This item is not returnable.
View return policy
Description
SIVA Polyclonal antibody specifically detects SIVA in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| SIVA | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Chicken | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 532 | |
| CD27-binding (Siva) protein, CD27BPCD27-binding protein, Siva-1, SIVA1, apoptosis-inducing factor, Siva-2, SIVAapoptosis regulatory protein Siva | |
| Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human SIVA. The immunogen is located within the first 50 amino acids of SIVA. Amino Acid Squence: AERYSQEVFEKTKRL | |
| 0.1 mL | |
| Apoptosis | |
| 10572 | |
| Store at 4°C in the dark. | |
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction