missing translation for 'onlineSavingsMsg'
Learn More

Sitravatinib, MedChemExpress

Product Code. p-200096769
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489747 5 mg 5mg
16477196 100 mg 100mg
16441236 200 mg 200mg
16407228 10 mg 10mg
16403247 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16489747 Supplier Medchem Express Supplier No. HY16961

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sitravatinib (MGCD516) is an orally bioavailable receptor tyrosine kinase (RTK) inhibitor with IC50s of 1.5 nM, 2 nM, 2 nM, 5 nM, 6 nM, 6 nM, 8 nM, 0.5 nM, 29 nM, 5 nM, and 9 nM for Axl, MER, VEGFR3, VEGFR2, VEGFR1, KIT, FLT3, DDR2, DDR1, TRKA, TRKB, respectively. Sitravatinib shows potent single-agent antitumor efficacy and enhances the activity of PD-1 blockade through promoting an antitumor immune microenvironment.

Sitravatinib (MGCD516) is an orally bioavailable receptor tyrosine kinase (RTK) inhibitor with IC50s of 1.5 nM, 2 nM, 2 nM, 5 nM, 6 nM, 6 nM, 8 nM, 0.5 nM, 29 nM, 5 nM, and 9 nM for Axl, MER, VEGFR3, VEGFR2, VEGFR1, KIT, FLT3, DDR2, DDR1, TRKA, TRKB, respectively. Sitravatinib shows potent single-agent antitumor efficacy and enhances the activity of PD-1 blockade through promoting an antitumor immune microenvironment.

Specifications

Chemical Name or Material Sitravatinib
CAS 1123837-84-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C33H29F2N5O4S
Quantity 5 mg
Synonym MGCD516 MG-516
Solubility Information DMSO : ≥ 32 mg/mL (50.82 mM)
SMILES O=C(C1(C(NC2=CC=C(F)C=C2)=O)CC1)NC3=CC=C(OC4=C5C(C=C(C6=NC=C(CNCCOC)C=C6)S5)=NC=C4)C(F)=C3
Molecular Weight (g/mol) 629.68
Formula Weight 629.68
Percent Purity 98.2%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.