missing translation for 'onlineSavingsMsg'
Learn More

SIT1 Antibody (SIT-01), FITC, Novus Biologicals™

Product Code. 18520573 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18520573 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18520573 Supplier Novus Biologicals Supplier No. NB500486F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SIT1 Monoclonal antibody specifically detects SIT1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Flow (Intracellular), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIT1
Applications Western Blot, Flow Cytometry, Immunoprecipitation, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone SIT-01
Conjugate FITC
Dilution Western Blot, Flow Cytometry, Immunoprecipitation, Flow (Intracellular), CyTOF-ready
Formulation PBS
Gene Alias gp30/40, MGC125908, MGC125909, RP11-331F9.5, SHP2 interacting transmembrane adaptor, SHP2-interacting transmembrane adapter protein, SHP2-interacting transmembrane adaptor protein, signaling threshold regulating transmembrane adaptor 1, signaling threshold-regulating transmembrane adapter 1, SITMGC125910, suppression inducing transmembrane adaptor 1, Suppression-inducing transmembrane adapter 1
Host Species Mouse
Immunogen Bacterially produced recombinant intracellular fragment of human SIT.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 27240
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.