missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIT1 Antibody (SIT-01), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-486B
This item is not returnable.
View return policy
Description
SIT1 Monoclonal antibody specifically detects SIT1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Flow (Intracellular), CyTOF
Specifications
| SIT1 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunoprecipitation, Flow Cytometry (Intracellular Staining), CyTOF | |
| SIT-01 | |
| Western Blot, Flow Cytometry, Immunoprecipitation, Flow (Intracellular), CyTOF-ready | |
| gp30/40, MGC125908, MGC125909, RP11-331F9.5, SHP2 interacting transmembrane adaptor, SHP2-interacting transmembrane adapter protein, SHP2-interacting transmembrane adaptor protein, signaling threshold regulating transmembrane adaptor 1, signaling threshold-regulating transmembrane adapter 1, SITMGC125910, suppression inducing transmembrane adaptor 1, Suppression-inducing transmembrane adapter 1 | |
| Bacterially produced recombinant intracellular fragment of human SIT. | |
| 0.1 mL | |
| Signal Transduction | |
| 27240 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction