missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIT1 Antibody (SIT-01), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-486AF700
This item is not returnable.
View return policy
Description
SIT1 Monoclonal specifically detects SIT1 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| SIT1 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| gp30/40, MGC125908, MGC125909, RP11-331F9.5, SHP2 interacting transmembrane adaptor, SHP2-interacting transmembrane adapter protein, SHP2-interacting transmembrane adaptor protein, signaling threshold regulating transmembrane adaptor 1, signaling threshold-regulating transmembrane adapter 1, SITMGC125910, suppression inducing transmembrane adaptor 1, Suppression-inducing transmembrane adapter 1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The antibody SIT-01 reacts with SHP2-interacting transmembrane adaptor protein (SIT) expressed exclusively in lymphoid organs. | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry | |
| SIT-01 | |
| Western Blot, Flow Cytometry | |
| SIT1 | |
| Bacterially produced recombinant intracellular fragment of human SIT. | |
| 0.1 mL | |
| Signal Transduction | |
| 27240 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction