missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIT1 Antibody (SIT-01), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-486AF594
This item is not returnable.
View return policy
Description
SIT1 Monoclonal specifically detects SIT1 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| SIT1 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SIT1 | |
| Bacterially produced recombinant intracellular fragment of human SIT. | |
| Primary | |
| The antibody SIT-01 reacts with SHP2-interacting transmembrane adaptor protein (SIT) expressed exclusively in lymphoid organs. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry | |
| SIT-01 | |
| Western Blot, Flow Cytometry | |
| gp30/40, MGC125908, MGC125909, RP11-331F9.5, SHP2 interacting transmembrane adaptor, SHP2-interacting transmembrane adapter protein, SHP2-interacting transmembrane adaptor protein, signaling threshold regulating transmembrane adaptor 1, signaling threshold-regulating transmembrane adapter 1, SITMGC125910, suppression inducing transmembrane adaptor 1, Suppression-inducing transmembrane adapter 1 | |
| Mouse | |
| 0.1 mL | |
| 27240 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction