missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIT1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-52153
This item is not returnable.
View return policy
Description
SIT1 Polyclonal specifically detects SIT1 in Human samples. It is validated for Peptide ELISA.
Specifications
| SIT1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| gp30/40, MGC125908, MGC125909, RP11-331F9.5, SHP2 interacting transmembrane adaptor, SHP2-interacting transmembrane adapter protein, SHP2-interacting transmembrane adaptor protein, signaling threshold regulating transmembrane adaptor 1, signaling threshold-regulating transmembrane adapter 1, SITMGC125910, suppression inducing transmembrane adaptor 1, Suppression-inducing transmembrane adapter 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| ELISA | |
| 0.5 mg/mL | |
| Peptide ELISA Detection limit 1:2000 | |
| NP_055265.1 | |
| SIT1 | |
| Peptide with sequence C-HLSQWTRGRSRSH corresponding to internal region according to NP_055265.1. | |
| 0.1 mg | |
| Signal Transduction | |
| 27240 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction