missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SIS3
GREENER_CHOICE

Product Code. 18772459
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18772459

Brand: Tocris Bioscience™ 5291/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective Smad3 inhibitor; inhibits TGF-βR1 signaling

SIS3 is a selective Smad3 inhibitor; attenuates TGF-β1-dependent Smad3 phosphorylation and DNA binding. Has no effect on Smad2, p38 MAPK, ERK or PI 3-kinase signaling. Inhibits TGF-β1-induced myofibroblast differentiation of dermal fibroblasts. Also inhibits TGF-β2-induced endothelial cell differentiation in iPSCs.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mg
Product Type SIS3
Purity 97%
Molecular Weight (g/mol) 489.99

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.