missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SIS17
GREENER_CHOICE

Product Code. p-200050992
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18064262

Brand: Tocris Bioscience™ 7969/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective HDAC11 inhibitor

SIS17 is a selective HDAC11 inhibitor (IC50 = 0.83 μM). Inhibits demyristoylation of serine hydroxymethyl transferase 2 with no detectable activity at other HDAC subtypes. Combination with Oxaliplatin (Cat. No. 2623) increases cytotoxicity to K562 cells; improves survival of Oxaliplatin-treated 769P cells. Cell permeable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N'-Hexadecylthiophene-2-carbohydrazide
CAS 2374313-54-7
Quantity 50 mg
Target HDAC11
Molecular Formula C21H38N2OS
Purity 0.95
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.