missing translation for 'onlineSavingsMsg'
Learn More

Sirtuin modulator 1, MedChemExpress

Product Code. p-200096141
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16497888

Brand: Medchem Express HY19758A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sirtuin modulator 1 is a modulator of SIRTl, a homolog of SIRT3, with EC1.5 of < 1 μM, extracted from patent WO 2010071853 A1, Compound No.4.

Sirtuin modulator 1 is a modulator of SIRTl, a homolog of SIRT3, with EC1.5 of < 1 μM, extracted from patent WO 2010071853 A1, Compound No.4.

Specifications

Chemical Name or Material Sirtuin modulator 1
Physical Form Solid
CAS 2070015-26-6
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C31H32ClN5O2S2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 12.5 mg/mL (20.62 mM; Need ultrasonic)
SMILES O=C(C1=C(CCCOC)SC(C2=CC=CC=C2)=N1)NC3=CC=CC=C3C4=NC5=C(N=CC(CN6CCCC6)=C5)S4.Cl[H]
Molecular Weight (g/mol) 606.2
Formula Weight 606.2
Percent Purity 99.72%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.