missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sirtuin modulator 1, MedChemExpress
Description
Sirtuin modulator 1 is a modulator of SIRTl, a homolog of SIRT3, with EC1.5 of < 1 μM, extracted from patent WO 2010071853 A1, Compound No.4.
Specifications
Specifications
| Chemical Name or Material | Sirtuin modulator 1 |
| Physical Form | Solid |
| CAS | 2070015-26-6 |
| Color | Light Yellow |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C31H32ClN5O2S2 |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | DMSO : 12.5 mg/mL (20.62 mM; Need ultrasonic) |
| SMILES | O=C(C1=C(CCCOC)SC(C2=CC=CC=C2)=N1)NC3=CC=CC=C3C4=NC5=C(N=CC(CN6CCCC6)=C5)S4.Cl[H] |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?