missing translation for 'onlineSavingsMsg'
Learn More

Sirtuin 5/SIRT5 Antibody, Novus Biologicals™

Product Code. 18425721 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425721 0.1 mL 0.1mL
18467640 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18425721 Supplier Novus Biologicals Supplier No. NBP186004

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Sirtuin 5/SIRT5 Polyclonal antibody specifically detects Sirtuin 5/SIRT5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sirtuin 5/SIRT5
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias EC 3.5.1, EC 3.5.1.-, FLJ36950, NAD-dependent deacetylase sirtuin-5, silent mating type information regulation 2, S.cerevisiae, homolog 5, SIR2L5, sir2-like 5, SIR2-like protein 5, sirtuin (silent mating type information regulation 2 homolog) 5 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S.cerevisiae, homolog) 5, sirtuin 5, sirtuin type 5
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: AKAKHIVIISGAGVSAESGVPTFRGAGGYWRKWQAQDLATPLAFAHNPSRVWEFYHYRREVMGSKEPNAGHRAIAECETRLGKQGRRVVVITQN
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 23408
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.