missing translation for 'onlineSavingsMsg'
Learn More

Sirtuin 3/SIRT3 Rabbit anti-Human, Mouse, Rat, Clone: 7O4G7, Novus Biologicals™

Product Code. 18370546
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370546 100 μg 100µL
18334932 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18370546 Supplier Novus Biologicals Supplier No. NBP316083100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Sirtuin 3/SIRT3 Monoclonal antibody specifically detects Sirtuin 3/SIRT3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sirtuin 3/SIRT3
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown
Classification Monoclonal
Clone 7O4G7
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias EC 3.5.1, EC 3.5.1.-, hSIRT3, mitochondrial nicotinamide adenine dinucleotide-dependent deacetylase, NAD-dependent deacetylase sirtuin-3, mitochondrial, SIR2L3silent mating type information regulation 2, S.cerevisiae, homolog 3, sir2-like 3, SIR2-like protein 3, sirtuin (silent mating type information regulation 2 homolog) 3 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S.cerevisiae, homolog) 3, sirtuin 3, sirtuin type 3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 300-399 of human Sirtuin 3/SIRT3 (NP_036371.1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 23410
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.