missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sirtuin 3/SIRT3 Antibody (PCRP-SIRT3-1C10), DyLight 755, Novus Biologicals™
Description
Sirtuin 3/SIRT3 Monoclonal antibody specifically detects Sirtuin 3/SIRT3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | Sirtuin 3/SIRT3 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | PCRP-SIRT3-1C10 |
| Conjugate | DyLight 755 |
| Dilution | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 3.5.1, EC 3.5.1.-, hSIRT3, mitochondrial nicotinamide adenine dinucleotide-dependent deacetylase, NAD-dependent deacetylase sirtuin-3, mitochondrial, SIR2L3silent mating type information regulation 2, S.cerevisiae, homolog 3, sir2-like 3, SIR2-like protein 3, sirtuin (silent mating type information regulation 2 homolog) 3 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S.cerevisiae, homolog) 3, sirtuin 3, sirtuin type 3 |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human Sirtuin 3/SIRT3 protein (Uniprot: Q9NTG7) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?