missing translation for 'onlineSavingsMsg'
Learn More

Sirtuin 3/SIRT3 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18421816 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421816 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421816 Supplier Novus Biologicals Supplier No. NBP176264MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chicken Polyclonal Antibody

Sirtuin 3/SIRT3 Polyclonal antibody specifically detects Sirtuin 3/SIRT3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sirtuin 3/SIRT3
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias EC 3.5.1, EC 3.5.1.-, hSIRT3, mitochondrial nicotinamide adenine dinucleotide-dependent deacetylase, NAD-dependent deacetylase sirtuin-3, mitochondrial, SIR2L3silent mating type information regulation 2, S.cerevisiae, homolog 3, sir2-like 3, SIR2-like protein 3, sirtuin (silent mating type information regulation 2 homolog) 3 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S.cerevisiae, homolog) 3, sirtuin 3, sirtuin type 3
Host Species Chicken
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the center of human SIRT3. The immunogen is located within amino acids 180 - 230 of SIRT3. Amino Acid Squence: LAKELYPGNYKPNVTHY
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 23410
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.