missing translation for 'onlineSavingsMsg'
Learn More

Sirtuin 3/SIRT3 Antibody [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30737074 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30737074 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737074 Supplier Novus Biologicals Supplier No. AF7488JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Sirtuin 3/SIRT3 Polyclonal antibody specifically detects Sirtuin 3/SIRT3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sirtuin 3/SIRT3
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias EC 3.5.1, EC 3.5.1.-, hSIRT3, mitochondrial nicotinamide adenine dinucleotide-dependent deacetylase, NAD-dependent deacetylase sirtuin-3, mitochondrial, SIR2L3silent mating type information regulation 2, S.cerevisiae, homolog 3, sir2-like 3, SIR2-like protein 3, sirtuin (silent mating type information regulation 2 homolog) 3 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S.cerevisiae, homolog) 3, sirtuin 3, sirtuin type 3
Host Species Sheep
Immunogen E. coli-derived recombinant human Sirtuin 3/SIRT3, Ser101-Lys399, Accession # Q9NTG7
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 23410
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.