missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sirtuin 2/SIRT2 Rabbit anti-Human, Clone: 1E1U3, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | Sirtuin 2/SIRT2 |
|---|---|
| Clone | 1E1U3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18333832
|
Bio-Techne
NBP3-16301-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18343785
|
Novus Biologicals
NBP3-16301-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sirtuin 2/SIRT2 Monoclonal antibody specifically detects Sirtuin 2/SIRT2 in Human samples. It is validated for Western BlotSpecifications
| Sirtuin 2/SIRT2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Chromatin Research, DNA Repair, Epigenetics, Neuronal Cell Markers, Transcription Factors and Regulators | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 22933 | |
| IgG | |
| Affinity purified |
| 1E1U3 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.5.1, EC 3.5.1.-, FLJ35621, FLJ37491, S.cerevisiae, homolog) 2, sirtuin 2, sirtuin type 2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Sirtuin 2/SIRT2 (Q8IXJ6). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel