missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sirtuin 2/SIRT2 Mouse anti-Human, Clone: PCRP-SIRT2-1A8, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | Sirtuin 2/SIRT2 |
|---|---|
| Clone | PCRP-SIRT2-1A8 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array |
Description
Sirtuin 2/SIRT2 Monoclonal specifically detects Sirtuin 2/SIRT2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.Specifications
| Sirtuin 2/SIRT2 | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Chromatin Research, DNA Repair, Epigenetics, Neuronal Cell Markers, Transcription Factors and Regulators | |
| 10 mM PBS with 0.05% BSA | |
| 22933 | |
| IgG1 | |
| Protein A or G purified |
| PCRP-SIRT2-1A8 | |
| Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.5.1, EC 3.5.1.-, FLJ35621, FLJ37491, S.cerevisiae, homolog) 2, sirtuin 2, sirtuin type 2 | |
| Recombinant full-length human Sirtuin 2/SIRT2 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title