missing translation for 'onlineSavingsMsg'
Learn More

Sirtuin 2/SIRT2 Antibody (PCRP-SIRT2-1A8) - Azide and BSA Free, Novus Biologicals™

Product Code. 18332663 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18332663 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18332663 Supplier Novus Biologicals Supplier No. NBP308293

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sirtuin 2/SIRT2 Monoclonal specifically detects Sirtuin 2/SIRT2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sirtuin 2/SIRT2
Applications Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array
Classification Monoclonal
Clone PCRP-SIRT2-1A8
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1-2 ug/mL, Flow Cytometry : 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence : 1-2 ug/mL, Protein Array
Formulation 10mM PBS
Gene Alias EC 3.5.1, EC 3.5.1.-, FLJ35621, FLJ37491, S.cerevisiae, homolog) 2, sirtuin 2, sirtuin type 2
Host Species Mouse
Immunogen Recombinant full-length human Sirtuin 2/SIRT2 protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Chromatin Research, DNA Repair, Epigenetics, Neuronal Cell Markers, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 22933
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.