missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Sirtuin 2/SIRT2 Polyclonal antibody specifically detects Sirtuin 2/SIRT2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Sirtuin 2/SIRT2 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 500 SE |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 3.5.1, EC 3.5.1.-, FLJ35621, FLJ37491, S.cerevisiae, homolog) 2, sirtuin 2, sirtuin type 2 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of the human SIRT2. The immunogen is located within the first 50 amino acids of SIRT2. Amino Acid Squence: AGKVQEAQDSDSDSEGG |
| Purification Method | Peptide affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?