missing translation for 'onlineSavingsMsg'
Learn More

Sirtuin 1/SIRT1 Antibody (PCRP-SIRT1-1E11), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18350474 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18350474 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18350474 Supplier Novus Biologicals Supplier No. NBP314290JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sirtuin 1/SIRT1 Monoclonal antibody specifically detects Sirtuin 1/SIRT1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sirtuin 1/SIRT1
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-SIRT1-1E11
Conjugate Janelia Fluor 549
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias EC 3.5.1, S. cerevisiae, homolog) 1, sirtuin 1
Host Species Mouse
Immunogen Recombinant full-length human Sirtuin 1/SIRT1protein (Uniprot: Q96EB6)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Chromatin Research, Cognition and Behavior, DNA Repair, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 23411
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.